Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide
Supplier: Apollo Scientific
Warning
Synonyms: Tetrahydro-2H-thiopyran-4-carboxylic acid 1,1-dioxide
OR301365-1G OR301365-5G OR301365-10G
APOSOR301365-1GEA 23.2 EUR
APOSOR301365-1G APOSOR301365-5G APOSOR301365-10G
Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide
Tetrahydro-2H-thiopyran-4-carboxylic acid-1,1-dioxide
Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
Melting Pt: 190…194 °C
Storage Temperature: Ambient
MDL Number: MFCD03102802
CAS Number: 64096-87-3