3-Amino-5-methyl-1H-pyrazole

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine

OR3015-100G OR3015-25G OR3015-5G
APOSOR3015-100GEA 125 EUR
APOSOR3015-100G APOSOR3015-25G APOSOR3015-5G
3-Amino-5-methyl-1H-pyrazole
3-Amino-5-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 213 °C/14mmHg
Melting Pt: 45…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR