Methyl-3-(chlorosulphonyl)thiophene-2-carboxylate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 3-(chlorosulphonyl)thiophene-2-carboxylate, 2-Carbomethoxy-3-thiophenesulphonyl chloride
OR3016-25G OR3016-5G
APOSOR3016-25GEA 43.5 EUR
APOSOR3016-25G APOSOR3016-5G
Methyl-3-(chlorosulphonyl)thiophene-2-carboxylate 97%
Methyl-3-(chlorosulphonyl)thiophene-2-carboxylate
Formula: C₆H₅ClO₄S₂
MW: 240.6845 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068160
CAS Number: 59337-92-7
UN: 3261
ADR: 8 III