4-Chloro-5-nitrophthalimide
Supplier: Apollo Scientific
|
Warning
|
OR30160-1G
OR30160-250MG
APOSOR30160-1GEA
116
EUR
APOSOR30160-1G
APOSOR30160-250MG
4-Chloro-5-nitrophthalimide
4-Chloro-5-nitrophthalimide
Formula:
C₈H₃ClN₂O₄ MW: 226.58 g/mol Melting Pt: 201…203 °C Density: 1.725 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00052331 CAS Number: 6015-57-2 |