3-Chlorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
|
Danger
|
OR30186-25G
OR30186-5G
APOSOR30186-25GEA
183
EUR
APOSOR30186-25G
APOSOR30186-5G
3-Chlorophenyl isothiocyanate 97%
3-Chlorophenyl isothiocyanate
Formula:
C₇H₄ClNS Boiling Pt: 249…250 °C Density: 1.281 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004805 CAS Number: 2392-68-9 UN: 2922 ADR: 8+6.1,II |