4-Methoxybenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
APOSOR30190-25GEA 17.6 EUR
APOSOR30190-25G APOSOR30190-250G APOSOR30190-100G
4-Methoxybenzenesulphonyl chloride 98%
4-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206,65 g/mol
Boiling Pt: 119…122 °C/0,4mmHg
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007446
EINECS: 202-692-1
UN: 3261
ADR: 8,II