Methyl acetopyruvate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2,4-dioxopentanoate
OR30191-25G
APOSOR30191-25GEA
75.4
EUR
APOSOR30191-25G
Methyl acetopyruvate 98%
Methyl acetopyruvate
Formula:
C₆H₈O₄ MW: 144.13 g/mol Melting Pt: 60…65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043638 CAS Number: 20577-61-1 EINECS: 243-891-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...