3'-Bromoacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: m-Bromoacetophenone
OR30202-100G OR30202-25G OR30202-500G
APOSOR30202-100GEA 65.4 EUR
APOSOR30202-100G APOSOR30202-25G APOSOR30202-500G
3'-Bromoacetophenone
3'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 79…81 °C/2mmHg
Melting Pt: 44508 °C
Density: 1.505 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4
EINECS: 218-396-0