1-Bromo-3-ethylbenzene 98%
Supplier: Apollo Scientific
Warning
OR30209-100G
APOSOR30209-100GEA 161 EUR
APOSOR30209-100G
1-Bromo-3-ethylbenzene 98%
1-Bromo-3-ethylbenzene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 76…78 °C/10mmHg
Density: 1.3493 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00156128
CAS Number: 2725-82-8