2-Chloro-4-iodotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-iodo-1-methylbenzene, 1-Chloro-5-iodo-2-methylbenzene
OR30229-100G OR30229-25G
APOSOR30229-100GEA 78.3 EUR
APOSOR30229-100G APOSOR30229-25G
2-Chloro-4-iodotoluene
2-Chloro-4-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 118…120 °C/15mmHg
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040872
CAS Number: 83846-48-4