1-Butyl isothiocyanate
Supplier: Apollo Scientific
Danger
Synonyms: Butyl isothiocyanate
OR30240-100G OR30240-25G OR30240-500G
APOSOR30240-100GEA 48 EUR
APOSOR30240-100G APOSOR30240-25G APOSOR30240-500G
1-Butyl isothiocyanate
1-Butyl isothiocyanate
Formula: C₅H₉NS
MW: 115.2 g/mol
Boiling Pt: 63…65 °C/11 mmHg
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004824
CAS Number: 592-82-5
EINECS: 209-770-4
UN: 2927
ADR: 6.1+8,II