N1-(4-Butyl-2-methylphenyl)acetamide
Supplier: Apollo Scientific
Warning
OR30289-1G OR30289-100MG OR30289-250MG OR30289-500MG OR30289-5G
APOSOR30289-1GEA 373 EUR
APOSOR30289-1G APOSOR30289-100MG APOSOR30289-250MG APOSOR30289-500MG APOSOR30289-5G
N1-(4-Butyl-2-methylphenyl)acetamide
N1-(4-Butyl-2-methylphenyl)acetamide
Formula: C₁₃H₁₉NO
Melting Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00277686
CAS Number: 143360-00-3