2,4-Dichlorothieno[3,2-d]pyrimidine 97%
Supplier: Apollo Scientific
Warning
OR303018-1G OR303018-5G
APOSOR303018-1GEA 21.9 EUR
APOSOR303018-1G APOSOR303018-5G
2,4-Dichlorothieno[3,2-d]pyrimidine 97%
2,4-Dichlorothieno[3,2-d]pyrimidine
Formula: C₆H₂Cl₂N₂S
MW: 205.0645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08448158
CAS Number: 16234-14-3