tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate
Supplier: Apollo Scientific
OR303048-1G OR303048-500MG
APOSOR303048-1GEA 538 EUR
APOSOR303048-1G APOSOR303048-500MG
tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate
tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate
Formula: C₁₅H₂₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19441845
CAS Number: 86499-69-6