Methyl-2-(4-hydroxy-2-methoxyphenyl)acetate
Supplier: Apollo Scientific
Warning
OR303108-1G OR303108-500MG
APOSOR303108-1GEA 1070 EUR
APOSOR303108-1G APOSOR303108-500MG
Methyl-2-(4-hydroxy-2-methoxyphenyl)acetate
Methyl-2-(4-hydroxy-2-methoxyphenyl)acetate
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399380
CAS Number: 499789-92-3