tert-Butyl-3-iodo-1H-indazole-1-carboxylate
Supplier: Apollo Scientific
Synonyms: tert-Butyl 3-iodo-1H-indazole-1-carboxylate
OR303121-1G OR303121-500MG
APOSOR303121-1GEA 180 EUR
APOSOR303121-1G APOSOR303121-500MG
tert-Butyl-3-iodo-1H-indazole-1-carboxylate
tert-Butyl-3-iodo-1H-indazole-1-carboxylate
Formula: C₁₂H₁₃IN₂O₂
MW: 344.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835520
CAS Number: 290368-00-2