4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Supplier: Apollo Scientific
OR303357-1G OR303357-5G OR303357-25G
APOSOR303357-1GEA 29 EUR
APOSOR303357-1G APOSOR303357-5G APOSOR303357-25G
4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
4,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999173
CAS Number: 115093-90-8