N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide
Supplier: Apollo Scientific
Synonyms: N-(6-Bromopyridin-2-yl)pivalamide
OR303633-1G OR303633-500MG
APOSOR303633-1GEA 270 EUR
APOSOR303633-1G APOSOR303633-500MG
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide
Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235018
CAS Number: 221087-47-4