5-Chloro-3-[2-(trimethylsilyl)ethynyl]-2-pyridinamine
Supplier: Apollo Scientific
Synonyms: 5-Chloro-3-((trimethylsilyl)ethynyl)pyridin-2-amine
OR303657-1G OR303657-500MG
APOSOR303657-1GEA 336 EUR
APOSOR303657-1G APOSOR303657-500MG
5-Chloro-3-[2-(trimethylsilyl)ethynyl]-2-pyridinamine
5-Chloro-3-[2-(trimethylsilyl)ethynyl]-2-pyridinamine
Formula: C₁₀H₁₃ClN₂Si
MW: 224.77 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD09880016
CAS Number: 866318-90-3