4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid
Supplier: Apollo Scientific
Synonyms: 4-Hydroxy-2-(pyridin-2-yl)pyrimidine-5-carboxylic acid
OR303680-1G OR303680-500MG
APOSOR303680-1GEA 336 EUR
APOSOR303680-1G APOSOR303680-500MG
4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid
4-Hydroxy-2-(2-pyridinyl)-5-pyrimidinecarboxylicacid
Formula: C₁₀H₇N₃O₃
MW: 217.19 g/mol
Melting Pt: 330 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11100252
CAS Number: 56406-45-2