(3-Chloropyridin-2-yl)methanol 95+%

Supplier: Apollo Scientific
Danger

OR303685-1G OR303685-250MG
APOSOR303685-1GEA 88.6 EUR
APOSOR303685-1G APOSOR303685-250MG
(3-Chloropyridin-2-yl)methanol 95+%
(3-Chloropyridin-2-yl)methanol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD11110226
CAS Number: 60588-81-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR