2-Chloro-6-cyclopropylnicotinonitrile
Supplier: Apollo Scientific
Warning
OR303703-1G OR303703-500MG
APOSOR303703-1GEA 95.7 EUR
APOSOR303703-1G APOSOR303703-500MG
2-Chloro-6-cyclopropylnicotinonitrile
2-Chloro-6-cyclopropylnicotinonitrile
Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD13193091
CAS Number: 1198475-35-2