6-(Phenylsulphanyl)-3-pyridinamine
Supplier: Apollo Scientific
OR303738-500MG
APOSOR303738-500MGEA
336
EUR
APOSOR303738-500MG
6-(Phenylsulphanyl)-3-pyridinamine
6-(Phenylsulphanyl)-3-pyridinamine
Formula:
C₁₁H₁₀N₂S MW: 202.28 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD03230019 CAS Number: 103983-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...