3,5-Dichloro-2-methoxyisonicotinic acid
Supplier: Apollo Scientific
Warning
OR303805-1G OR303805-500MG
APOSOR303805-1GEA 671 EUR
APOSOR303805-1G APOSOR303805-500MG
3,5-Dichloro-2-methoxyisonicotinic acid
3,5-Dichloro-2-methoxyisonicotinic acid
Formula: C₇H₅Cl₂NO₃
MW: 222.025 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD12025889
CAS Number: 1147979-40-5