2,5-Dichloro-4-formylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Dichloroisonicotinaldehyde
OR303891-5G
APOSOR303891-5GEA 97.3 EUR
APOSOR303891-5G
2,5-Dichloro-4-formylpyridine 97%
2,5-Dichloro-4-formylpyridine
Formula: C₆H₃Cl₂NO
MW: 176.0000 g/mol
Melting Pt: 73…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06410679
CAS Number: 102645-33-0