2-Chloro-3-methylquinoxaline
Supplier: Apollo Scientific
OR303925-1G OR303925-5G
APOSOR303925-1GEA 243 EUR
APOSOR303925-1G APOSOR303925-5G
2-Chloro-3-methylquinoxaline
2-Chloro-3-methylquinoxaline
Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00023913
CAS Number: 32601-86-8