(R)-(-)-Hydratropic acid 98%
Supplier: Apollo Scientific
OR304085-25G OR304085-5G
APOSOR304085-25GEA 136 EUR
APOSOR304085-25G APOSOR304085-5G
(R)-(-)-Hydratropic acid 98%
(R)-(-)-Hydratropic acid
Formula: C₉H₁₀O₂
Melting Pt: 29…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00063140
CAS Number: 7782-26-5