trans-3-(3-Pyridyl)acrylic acid 98%
Supplier: Apollo Scientific
Warning
OR30410-100G OR30410-250G OR30410-25G
APOSOR30410-100GEA 166 EUR
APOSOR30410-100G APOSOR30410-250G APOSOR30410-25G
trans-3-(3-Pyridyl)acrylic acid 98%
trans-3-(3-Pyridyl)acrylic acid
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 232…235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006410
CAS Number: 19337-97-4
EINECS: 214-424-0