2,2,6,6-Tetramethylpiperidine 99%
Supplier: Apollo Scientific
Danger
OR30412-100G OR30412-25G OR30412-500G
APOSOR30412-100GEA 43.5 EUR
APOSOR30412-100G APOSOR30412-25G APOSOR30412-500G
2,2,6,6-Tetramethylpiperidine 99%
2,2,6,6-Tetramethylpiperidine
Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 151…152 °C
Melting Pt: –59 °C
Density: 0.837 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005985
CAS Number: 768-66-1
EINECS: 212-199-3
UN: 1992
ADR: 3+6.1,III