5-Amino-1H-indazole 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine
OR30440-100G OR30440-5G OR30440-25G
APOSOR30440-100GEA 219 EUR
APOSOR30440-100G APOSOR30440-5G APOSOR30440-25G
5-Amino-1H-indazole 95%
5-Amino-1H-indazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 172…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00037975
CAS Number: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: 6.1,III