9-Oxa-2-azaspiro[5.5]undecane
Supplier: Apollo Scientific
OR304433-1G OR304433-5G
APOSOR304433-1GEA 521 EUR
APOSOR304433-1G APOSOR304433-5G
9-Oxa-2-azaspiro[5.5]undecane
9-Oxa-2-azaspiro[5.5]undecane
Formula: C₉H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD12827461
CAS Number: 57289-77-7