4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98% free radical
Supplier: Apollo Scientific
Warning
OR30449-25G OR30449-5G OR30449-100G
APOSOR30449-25GEA 139 EUR
APOSOR30449-25G APOSOR30449-5G APOSOR30449-100G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Formula: C₉H₁₉N₂O
Boiling Pt: 98 °C/3.75mmHg
Melting Pt: 33…39 °C
Density: 0.896 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2