Cyclobutanone 98%
Supplier: Apollo Scientific
|
Danger
|
OR30488-25G
OR30488-5G
OR30488-100G
APOSOR30488-25GEA
49.3
EUR
APOSOR30488-25G
APOSOR30488-5G
APOSOR30488-100G
Cyclobutanone 98%
Cyclobutanone
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 97…99 °C Melting Pt: –51 °C Density: 0.938 g/cm³ (25 °C) Flash Pt: 10 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001332 CAS Number: 1191-95-3 EINECS: 214-745-6 UN: 1224 ADR: 3 II |