3-Hydroxypropiononitrile
Supplier: Apollo Scientific
Warning
Synonyms: Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile
OR3049-1KG OR3049-250G
APOSOR3049-1KGEA 201 EUR
APOSOR3049-1KG APOSOR3049-250G
3-Hydroxypropiononitrile
3-Hydroxypropiononitrile
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 227…228 °C
Melting Pt: –46 °C
Density: 1.041 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828