5-Methyl-1H-indazole
Supplier: Apollo Scientific
Warning
OR305086-1G OR305086-5G OR305086-10G
APOSOR305086-1GEA 24.8 EUR
APOSOR305086-1G APOSOR305086-5G APOSOR305086-10G
5-Methyl-1H-indazole
5-Methyl-1H-indazole
Formula: C₈H₈N₂
MW: 132.1600 g/mol
Boiling Pt: 165…190 °C/1.5mmHg
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD01606235
CAS Number: 1776-37-0