Imidazo[1,2-a]pyridine-3-sulphonate
Supplier: Apollo Scientific
OR305156-250MG OR305156-1G
APOSOR305156-250MGEA 223 EUR
APOSOR305156-250MG APOSOR305156-1G
Imidazo[1,2-a]pyridine-3-sulphonate
Imidazo[1,2-a]pyridine-3-sulphonate
Formula: C₇H₆N₂O₃S
MW: 198.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176742
CAS Number: 112581-51-8