(±)-3-Methoxy-1,2-propanediol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methoxy-1,2-propanediol, (±)-3-Methoxy-1,2-dihydroxy propane
OR30517-100G OR30517-25G OR30517-5G
APOSOR30517-100GEA 395 EUR
APOSOR30517-100G APOSOR30517-25G APOSOR30517-5G
(±)-3-Methoxy-1,2-propanediol 98%
(±)-3-Methoxy-1,2-propanediol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 218…220 °C
Density: 1.114 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004719
CAS Number: 623-39-2
EINECS: 210-791-6