6-Methoxyisoindolin-1-one
Supplier: Apollo Scientific
|
Warning
|
OR305281-1G
OR305281-250MG
APOSOR305281-1GEA
87
EUR
APOSOR305281-1G
APOSOR305281-250MG
6-Methoxyisoindolin-1-one
6-Methoxyisoindolin-1-one
Formula:
C₉H₉NO₂ MW: 163.1733 g/mol Melting Pt: 189…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD08234735 CAS Number: 132680-54-7 |