5-Hydroxyisoindolin-1-one
Supplier: Apollo Scientific
Warning
OR305282-250MG OR305282-1G
APOSOR305282-250MGEA 72.5 EUR
APOSOR305282-250MG APOSOR305282-1G
5-Hydroxyisoindolin-1-one
5-Hydroxyisoindolin-1-one
Formula: C8H7NO2
MW: 149.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000830
CAS Number: 252061-66-8