6-Aminoisoindolin-1-one
Supplier: Apollo Scientific
Warning
Synonyms: 6-Amino-2,3-dihydro-1H-isoindol-1-one
OR305288-1G OR305288-5G
APOSOR305288-1GEA 108 EUR
APOSOR305288-1G APOSOR305288-5G
6-Aminoisoindolin-1-one
6-Aminoisoindolin-1-one
Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 247 °C
Storage Temperature: Ambient
MDL Number: MFCD09701282
CAS Number: 675109-45-2