6-Nitro-1-indanone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Nitro-2,3-dihydro-1H-inden-1-one
OR30531-1G
OR30531-5G
APOSOR30531-1GEA
27.7
EUR
APOSOR30531-1G
APOSOR30531-5G
6-Nitro-1-indanone 97%
6-Nitro-1-indanone
Formula:
C₉H₇NO₃ MW: 177.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656903 CAS Number: 24623-24-3 |