1-Methyl-3,5-dinitro-1H-pyridin-2-one 99%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Methyl-3,5-dinitro-2-pyridone, 1-Methyl-3,5-dinitropyridin-2(1H)-one
OR305320-1G OR305320-5G
APOSOR305320-1GEA 21.9 EUR
APOSOR305320-1G APOSOR305320-5G
1-Methyl-3,5-dinitro-1H-pyridin-2-one 99%
1-Methyl-3,5-dinitro-1H-pyridin-2-one
Formula: C₆H₅N₃O₅
MW: 199.121 g/mol
Melting Pt: 174…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00456280
CAS Number: 14150-94-8
UN: 2811
ADR: 6.1,III