6-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
Supplier: Apollo Scientific
Warning
OR305338-5G
APOSOR305338-5GEA 71.2 EUR
APOSOR305338-5G
6-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
6-Chloro-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17010105
CAS Number: 1206979-33-0