4-Amino-2-chloro-5-methylpyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-5-methylpyridin-4-amine
OR305585-1G
OR305585-5G
OR305585-25G
APOSOR305585-1GEA
21.9
EUR
APOSOR305585-1G
APOSOR305585-5G
APOSOR305585-25G
4-Amino-2-chloro-5-methylpyridine 98%
4-Amino-2-chloro-5-methylpyridine
Formula:
C₆H₇ClN₂ MW: 142.5862 g/mol Melting Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD11977435 CAS Number: 79055-62-2 |