1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
Supplier: Apollo Scientific
Warning
OR305629-1G OR305629-250MG
APOSOR305629-1GEA 310 EUR
APOSOR305629-1G APOSOR305629-250MG
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
Formula: C₈H₅N₃
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD09909831
CAS Number: 146767-63-7