5-Hexynoic acid 99%
Supplier: Apollo Scientific
Danger
OR30593-25G OR30593-1G OR30593-5G
APOSOR30593-25GEA 261 EUR
APOSOR30593-25G APOSOR30593-1G APOSOR30593-5G
5-Hexynoic acid 99%
5-Hexynoic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 224…225 °C
Density: 1.016 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 3265
ADR: 8,III