4-Bromo-o-cresol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene
OR3060-100G OR3060-25G OR3060-1G
APOSOR3060-100GEA 186 EUR
APOSOR3060-100G APOSOR3060-25G APOSOR3060-1G
4-Bromo-o-cresol 95%
4-Bromo-o-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Melting Pt: 62…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00055435
CAS Number: 2362-12-1