5-Bromo-1-pentene
Supplier: Apollo Scientific
Warning
OR30608-25G OR30608-5G
APOSOR30608-25GEA 48 EUR
APOSOR30608-25G APOSOR30608-5G
5-Bromo-1-pentene
5-Bromo-1-pentene
Formula: C₅H₉Br
Boiling Pt: 126…127 °C
Density: 1.258 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000264
CAS Number: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III