1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride
Supplier: Apollo Scientific
Warning
OR30668-250MG OR30668-1G
APOSOR30668-250MGEA 174 EUR
APOSOR30668-250MG APOSOR30668-1G
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride
Formula: C₈H₁₄ClNO₂
MW: 191.6600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869757
CAS Number: 374794-92-0