4-Bromo-3-nitrotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-methyl-2-nitrobenzene
OR3068-100G OR3068-25G
APOSOR3068-100GEA 74.1 EUR
APOSOR3068-100G APOSOR3068-25G
4-Bromo-3-nitrotoluene 99%
4-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 152 °C/14mmHg
Melting Pt: 31…33 °C
Density: 1.578 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6